2-Chloro-6-methoxy-3-nitropyridine ≥95%

Supplier: Matrix Scientific

073230-100G
102529-072EA 347.69 USD
102529-072
2-Chloro-6-methoxy-3-nitropyridine ≥95%
2-Chloro-6-methoxy-3-nitropyridine

Hazard: Irritant

Formula: C₆H₅ClN₂O₃
MW: 188.57 g/mol
MDL Number: MFCD00130268
CAS Number: 38533-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR