Methyl-6-bromopicolinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 6-bromopicolinateMethyl 6-bromopyridine-2-carboxylate6-Bromopyridine-2-carboxylic acid methyl ester6-Bromopicolinic acid methyl ester

073228-5G 073228-25G
102663-940EA 215.38 USD
102663-940 102529-060
Methyl-6-bromopicolinate ≥95%
Methyl 6-bromopicolinate

Hazard: Irritant/Light Sensitive

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 92…96 °C
MDL Number: MFCD06203934
CAS Number: 26218-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR