(2-Methyl-3-nitrophenyl)acetic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Methyl-3-nitrophenylacetic Acid
073222-10G
073222-5G
073222-100G
102529-028EA
46.15
USD
102529-028
102529-032
102529-030
(2-Methyl-3-nitrophenyl)acetic acid ≥95%
(2-Methyl-3-nitrophenyl)acetic acid
Hazard: Irritant
Formula:
C₉H₉NO₄ MW: 195.17 g/mol |
MDL Number:
MFCD07782132 CAS Number: 23876-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...