6-Bromo-3-isoquinolinol ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-3-hydroxyisoquinoline

073146-1G 073146-250MG
102528-626EA 347.56 USD
102528-626 102528-628
6-Bromo-3-isoquinolinol ≥95%
6-Bromo-3-isoquinolinol

Hazard: Irritant

Formula: C₉H₆BrNO
MW: 224.0500 g/mol
MDL Number: MFCD11110423
CAS Number: 1031927-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR