Methyl-2-fluoro-3-nitrobenzoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-fluoro-3-nitrobenzoate
073122-5G
073122-1G
102528-496EA
1720
USD
102528-496
102528-494
Methyl-2-fluoro-3-nitrobenzoate ≥95%
Methyl 2-fluoro-3-nitrobenzoate
Hazard: Irritant
Formula:
C₈H₆FNO₄ MW: 199.1400 g/mol |
MDL Number:
MFCD10566813 CAS Number: 946126-94-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...