2-Fluoro-3-nitrobenzylalcohol ≥95%
Supplier: Matrix Scientific
Synonyms:
(2-Fluoro-3-nitrophenyl)methanol
073120-1G
073120-5G
073120-250MG
102528-486EA
2061.54
USD
102528-486
102528-482
102528-484
2-Fluoro-3-nitrobenzylalcohol ≥95%
2-Fluoro-3-nitrobenzylalkohol
Hazard: Irritant
Formula:
C₇H₆FNO₃ MW: 171.1300 g/mol |
MDL Number:
MFCD09835208 CAS Number: 946126-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...