5-Nitro-2-thiophenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Nitrothiophene-2-carboxylic acid

073115-5G 073115-1G 073115-10G
102528-456EA 51.75 USD
102528-456 102528-458 102528-454
5-Nitro-2-thiophenecarboxylic acid ≥95%
5-Nitro-2-thiophenecarboxylic acid

Hazard: Irritant

Formula: C₅H₃NO₄S
MW: 173.14 g/mol
MDL Number: MFCD00159552
CAS Number: 6317-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR