1,2,3,4-Tetrahydroquinoline ≥95%

Supplier: Matrix Scientific

073083-100G
102528-302EA 143.08 USD
102528-302
1,2,3,4-Tetrahydroquinoline ≥95%
1,2,3,4-Tetrahydroquinoline

Hazard: Irritant

Formula: C₉H₁₁N
MDL Number: MFCD00006693
CAS Number: 635-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR