Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloroanthranilic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-chlorobenzoic acid

073076-100G
102528-270EA 90.23 USD InStock
102528-270
5-Chloroanthranilic acid ≥95%
5-Chloroanthranilic acid
Purity: 95+%
CAS: 635-21-2
MDL Number: MFCD00007838
Molecular Formula: C7H6ClNO2
Molecular Weight: 171,58
Hazard: Irritant


Formula: C₇H₆ClNO₂
MW: 171,58 g/mol
Storage Temperature: ambient
MDL Number: MFCD00007838
CAS Number: 635-21-2
EINECS: 211-230-8
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty