5-Chloro-2-nitroaniline ≥95%

Supplier: Matrix Scientific

073071-100G
102528-246EA 67.69 USD
102528-246
5-Chloro-2-nitroaniline ≥95%
5-Chloro-2-nitroaniline

Hazard: Irritant

Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
MDL Number: MFCD00007776
CAS Number: 1635-61-6
UN: 2237
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR