2-Chloro-4-hydroxypyridine ≥95%

Supplier: Matrix Scientific

073070-5G
102528-240EA 64.62 USD
102528-240
2-Chloro-4-hydroxypyridine ≥95%
2-Chloro-4-hydroxypyridine

Hazard: Irritant

Formula: C₅H₄ClNO
MW: 129.55 g/mol
Melting Pt: 169…172 °C
MDL Number: MFCD01646108
CAS Number: 17368-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR