3-Amino-4-bromo-6-chloropyridazine ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Bromo-6-chloro-pyridazin-3-amine
073049-5G
102528-132EA
112.31
USD
102528-132
3-Amino-4-bromo-6-chloropyridazine ≥95%
3-Amino-4-bromo-6-chloropyridazine
Hazard: Irritant
Formula:
C₄H₃BrClN₃ MW: 208.4437 g/mol |
MDL Number:
MFCD11520891 CAS Number: 446273-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...