1-Aminocyclopropanecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Amino-1-cyclopropanecarboxylic acid

073019-5G
102527-956EA 84.62 USD
102527-956
1-Aminocyclopropanecarboxylic acid ≥95%
1-Aminocyclopropanecarboxylic acid

Hazard: Irritant

Formula: C₄H₇NO₂
MW: 101.11 g/mol
MDL Number: MFCD00009944
CAS Number: 22059-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR