6-Bromooxindole ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromoindolin-2-one6-Bromo-2-oxindole

072979-5G
102527-754EA 110.77 USD
102527-754
6-Bromooxindole ≥95%
6-Bromooxindole

Hazard: Irritant-Harmful/Stored under Argon/Light Sensitive

Formula: C₈H₆BrNO
MW: 212.05 g/mol
Melting Pt: 217…221 °C
MDL Number: MFCD02179605
CAS Number: 99365-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR