2-Chloro-6-nitroaniline ≥95%
Supplier: Matrix Scientific
072945-5G
072945-1G
072945-10G
102527-580EA
427.69
USD
102527-580
102527-584
102527-582
2-Chloro-6-nitroaniline ≥95%
2-Chloro-6-nitroaniline
Hazard: Irritant
Formula:
C₆H₅ClN₂O₂ MW: 172.57 g/mol |
MDL Number:
MFCD00179568 CAS Number: 769-11-9 UN: 2237 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...