7-Methoxy-1-tetralone ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Methoxy-3,4-dihydronaphthalen-1(2H)-one

072928-100G
102527-494EA 152.31 USD
102527-494
7-Methoxy-1-tetralone ≥95%
7-Methoxy-1-tetralone

Hazard: Irritant

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Melting Pt: 59…63 °C
MDL Number: MFCD00001696
CAS Number: 6836-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR