2-Bromo-3-nitroanisole ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-1-methoxy-3-nitrobenzene

072927-5G
102527-486EA 56.92 USD
102527-486
2-Bromo-3-nitroanisole ≥95%
2-Bromo-3-nitroanisole

Hazard: Irritant/Light Sensitive

Formula: C₇H₆BrNO₃
MW: 232.0300 g/mol
Melting Pt: 93…104 °C
MDL Number: MFCD07779272
CAS Number: 67853-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR