6-Bromo-2-pyridineboronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine6-Bromo-2-pyridylboronic acid Pinacol ester6-Bromopyridine-2-boronic acid pinacol ester2-(6-Bromo-2-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

072917-5G
102527-434EA 196.92 USD
102527-434
6-Bromo-2-pyridineboronic acid pinacol ester ≥95%
6-Bromo-2-pyridineboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₁H₁₅BBrNO₂
MW: 283.96 g/mol
MDL Number: MFCD06796177
CAS Number: 651358-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR