5-Nitroisoquinoline ≥95%

Supplier: Matrix Scientific

072905-10G 072905-100G
102527-374EA 67.85 USD
102527-374 102527-372
5-Nitroisoquinoline ≥95%
5-Nitroisoquinoline

Hazard: Irritant

Formula: C₉H₆N₂O₂
MDL Number: MFCD00006905
CAS Number: 607-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR