2,4-Diamino-6-hydroxypyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 2,4-Diamino-6-pyrimidinol6-Aminoisocytosin2,4-Diaminopyrimidin-6-ol

072892-100G
102527-302EA 135.38 USD
102527-302
2,4-Diamino-6-hydroxypyrimidine ≥95%
2,4-Diamino-6-hydroxypyrimidine

Hazard: Irritant

Formula: C₄H₆N₄O
MW: 126.12 g/mol
MDL Number: MFCD00006098
CAS Number: 56-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR