3-Amino-5-bromobenzonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-3-bromobenzonitrile

072870-1G
102527-192EA 67.85 USD
102527-192
3-Amino-5-bromobenzonitrile ≥95%
3-Amino-5-bromobenzonitrile

Hazard: Toxic/Refrigerate

Formula: C₇H₅BrN₂
MW: 197.0300 g/mol
Melting Pt: 124…125 °C
MDL Number: MFCD11109874
CAS Number: 49674-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR