2-Fluoro-5-nitropyridine ≥95%

Supplier: Matrix Scientific

072864-5G
102527-160EA 34.5 USD
102527-160
2-Fluoro-5-nitropyridine ≥95%
2-Fluoro-5-nitropyridine

Hazard: Irritant

Formula: C₅H₃FN₂O₂
MDL Number: MFCD03095059
CAS Number: 456-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR