2-Bromo-3-fluorophenol ≥95%

072862-5G
102527-148EA 124.52 USD
102527-148
2-Bromo-3-fluorophenol ≥95%
2-Bromo-3-fluorophenol

Hazard: Irritant/Refrigerate

Formula: C₆H₄BrFO
MW: 190.9978 g/mol
MDL Number: MFCD04037220
CAS Number: 443-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR