Methyl-1H-indazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 1H-indazole-3-carboxylate

072855-1G 072855-5G
102527-108EA 15.38 USD
102527-108 102527-110
Methyl-1H-indazole-3-carboxylate ≥95%
Methyl 1H-indazole-3-carboxylate

Hazard: Irritant

Formula: C₉H₈N₂O₂
MW: 176.1700 g/mol
MDL Number: MFCD01138133
CAS Number: 43120-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR