2,6-Dichloronicotinonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Dichloro-3-cyanopyridine2,6-Dichloropyridine-3-carbonitrile

072847-5G
102527-066EA 57.5 USD
102527-066
2,6-Dichloronicotinonitrile ≥95%
2,6-Dichloronicotinonitrile

Hazard: Irritant

Formula: C₆H₂Cl₂N₂
MW: 173.00 g/mol
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR