Isonipecotamide ≥95%

Supplier: Matrix Scientific

Synonyms: Piperidine-4-carboxamide

072839-5G 072839-10G 072839-100G
102527-026EA 413.85 USD
102527-026 102527-024 102527-022
Isonipecotamide ≥95%
Isonipecotamide

Hazard: Irritant

Formula: C₆H₁₂N₂O
MW: 128.18 g/mol
MDL Number: MFCD00038012
CAS Number: 39546-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR