2-Amino-4'-methoxyacetophenone hydrochloride ≥95%

Supplier: Matrix Scientific

072836-10G 072836-5G 072836-1G
102527-008EA 719.9 USD
102527-008 102527-006 102527-004
2-Amino-4'-methoxyacetophenone hydrochloride ≥95%
2-Amino-4'-methoxyacetophenone hydrochloride

Hazard: Irritant

Formula: C₉H₁₂ClNO₂
MDL Number: MFCD00193078
CAS Number: 3883-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR