5-Amino-2-bromo-3-methylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-2-bromo-3-picoline

072833-5G
102526-990EA 118.45 USD
102526-990
5-Amino-2-bromo-3-methylpyridine ≥95%
5-Amino-2-bromo-3-methylpyridine

Hazard: Irritant

Formula: C₆H₇BrN₂
MW: 187.04 g/mol
MDL Number: MFCD03095085
CAS Number: 38186-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR