2-Fluoro-4-hydroxybenzaldehyde ≥95%
Supplier: Matrix Scientific
072823-5G
102526-940EA
106.15
USD
102526-940
2-Fluoro-4-hydroxybenzaldehyde ≥95%
2-Fluoro-4-hydroxybenzaldehyde
Hazard: Irritant
Formula:
C₇H₅FO₂ MW: 140.11 g/mol |
MDL Number:
MFCD06797918 CAS Number: 348-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...