6-Fluoro-1H-indazole ≥95%

Supplier: Matrix Scientific

072822-1G 072822-5G 072822-10G
102526-932EA 146.15 USD
102526-932 102526-934 102526-936
6-Fluoro-1H-indazole ≥95%
6-Fluoro-1H-indazole

Hazard: Irritant

Formula: C₇H₅FN₂
MW: 136.1264 g/mol
MDL Number: MFCD07371562
CAS Number: 348-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR