5-Bromo-2-chlorobenzaldehyde ≥95%

Supplier: Matrix Scientific

072756-25G 072756-5G
102526-572EA 50.77 USD
102526-572 102833-546
5-Bromo-2-chlorobenzaldehyde ≥95%
5-Bromo-2-chlorobenzaldehyde

Hazard: Irritant

Formula: C₇H₄BrClO
MW: 219.46 g/mol
MDL Number: MFCD08445659
CAS Number: 189628-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR