1-Boc-4-(methylamino)piperidine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Methylamino)piperidine, N1-BOC protectedtert-Butyl 4-(methylamino)piperidine-1-carboxylate

072731-5G
102526-436EA 78.46 USD
102526-436
1-Boc-4-(methylamino)piperidine ≥95%
1-Boc-4-(methylamino)piperidine

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MW: 214.3046 g/mol
MDL Number: MFCD02259411
CAS Number: 147539-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR