2-Methyl-10H-benzo[b]thieno[2,3-e][1,4]diazepin-4-amine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-2-methyl-10H-thieno[2,3-b][1,5]benzodiazepine hydrochloride

072724-5G
102526-394EA 269.23 USD
102526-394
2-Methyl-10H-benzo[b]thieno[2,3-e][1,4]diazepin-4-amine hydrochloride ≥95%
2-Methyl-10H-benzo[b]thieno[2,3-e][1,4]diazepin-4-amine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₂ClN₃S
MW: 265.7618 g/mol
MDL Number: MFCD03411581
CAS Number: 138564-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR