4-Fluoro-2-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

072657-5G
102526-034EA 104.62 USD
102526-034
4-Fluoro-2-nitrobenzaldehyde ≥95%
4-Fluoro-2-nitrobenzaldehyde

Hazard: Irritant

Formula: C₇H₄FNO₃
MDL Number: MFCD01666439
CAS Number: 2923-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR