4-Hydroxy-2,5-dimethylbenzaldehyde ≥95%

Supplier: Matrix Scientific

072564-5G
102525-538EA 258.46 USD
102525-538
4-Hydroxy-2,5-dimethylbenzaldehyde ≥95%
4-Hydroxy-2,5-dimethylbenzaldehyde

Hazard: Irritant

Formula: C₉H₁₀O₂
MW: 150.1745 g/mol
MDL Number: MFCD03411744
CAS Number: 85231-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR