5-Isoquinolinol ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Hydroxyisoquinoline

072532-5G 072532-25G
102525-352EA 143.08 USD
102525-352 102833-534
5-Isoquinolinol ≥95%
5-Isoquinolinol

Hazard: Irritant

Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 231…233 °C
MDL Number: MFCD00006906
CAS Number: 2439-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR