4-Hydroxy-2-methylbenzaldehyde ≥95%

Supplier: Matrix Scientific

072527-5G
102525-324EA 196.92 USD
102525-324
4-Hydroxy-2-methylbenzaldehyde ≥95%
4-Hydroxy-2-methylbenzaldehyde

Hazard: Irritant

Formula: C₈H₈O₂
MW: 136.1479 g/mol
MDL Number: MFCD04037443
CAS Number: 41438-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR