2-Aminoquinazoline ≥95%

Supplier: Matrix Scientific

072499-5G 072499-1G
102525-168EA 1150.77 USD
102525-168 102525-166
2-Aminoquinazoline ≥95%
2-Aminoquinazoline

Hazard: Irritant

Formula: C₈H₇N₃
MW: 145.1600 g/mol
MDL Number: MFCD00956186
CAS Number: 1687-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR