4-Bromo-1-naphthol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromonaphthalen-1-ol

072433-5G 072433-1G
102524-816EA 53.85 USD
102524-816 102524-818
4-Bromo-1-naphthol ≥95%
4-Bromo-1-naphthol

Hazard: Irritant

Formula: C₁₀H₇BrO
MW: 223.0700 g/mol
MDL Number: MFCD03129265
CAS Number: 571-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR