1,3-Dibromoisoquinoline ≥95%
Supplier: Matrix Scientific
072423-5G
072423-250MG
072423-1G
102524-764EA
284.76
USD
102524-764
102524-766
102524-768
1,3-Dibromoisoquinoline ≥95%
1,3-Dibromoisoquinoline
Hazard: Irritant
Formula:
C₉H₅Br₂N MW: 286.9500 g/mol |
MDL Number:
MFCD00234474 CAS Number: 53987-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...