6-Bromosalicylaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Bromo-6-hydroxybenzaldehyde
072390-5G
102524-600EA
223.08
USD
102524-600
6-Bromosalicylaldehyde ≥95%
6-Bromosalicylaldehyde
Hazard: Irritant/Light Sensitive/Refrigerate
Formula:
C₇H₅BrO₂ MW: 201.0200 g/mol Melting Pt: 51…52 °C |
MDL Number:
MFCD08234722 CAS Number: 22532-61-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...