6-Methylpurine ≥95%

Supplier: Matrix Scientific

072386-250MG 072386-5G 072386-1G
102524-578EA 2061.54 USD
102524-578 102524-574 102524-576
6-Methylpurine ≥95%
6-Methylpurine

TCA listed: T
Hazard: Irritant

Formula: C₆H₆N₄
MW: 134.1400 g/mol
Melting Pt: 235…236 °C
MDL Number: MFCD00083663
CAS Number: 2004-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR