2-Amino-N,N-diethylbenzenesulfonamide ≥95%

Supplier: Matrix Scientific

072360-1G 072360-5G
102524-436EA 399.05 USD
102524-436 102524-438
2-Amino-N,N-diethylbenzenesulfonamide ≥95%
2-Amino-N,N-diethylbenzenesulphonamide

Hazard: Irritant

Formula: C₁₀H₁₆N₂O₂S
MDL Number: MFCD09736559
CAS Number: 57947-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR