2-Amino-N-propylbenzenesulfonamide ≥95%

Supplier: Matrix Scientific

072358-5G 072358-1G
102524-426EA 569.23 USD
102524-426 102524-428
2-Amino-N-propylbenzenesulfonamide ≥95%
2-Amino-N-propylbenzenesulphonamide

Hazard: Irritant

Formula: C₉H₁₄N₂O₂S
MDL Number: MFCD04035389
CAS Number: 436095-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR