2-Amino-N-isopropylbenzenesulfonamide ≥95%

Supplier: Matrix Scientific

072357-5G
102524-424EA 1535.38 USD
102524-424
2-Amino-N-isopropylbenzenesulfonamide ≥95%
2-Amino-N-isopropylbenzenesulphonamide

Hazard: Irritant

Formula: C₉H₁₄N₂O₂S
MW: 214.2900 g/mol
MDL Number: MFCD09730175
CAS Number: 761435-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR