5-Bromo-2-pyrazinecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromopyrazine-2-carboxylic acid
072316-1G
072316-5G
102524-214EA
295.38
USD
102524-214
102524-212
5-Bromo-2-pyrazinecarboxylic acid ≥95%
5-Bromo-2-pyrazinecarboxylic acid
Hazard: Irritant
Formula:
C₅H₃BrN₂O₂ MW: 203.00 g/mol |
MDL Number:
MFCD09909672 CAS Number: 876161-05-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...