2-Amino-5-bromobenzothiazole ≥95%
Supplier: Matrix Scientific
072271-1G
072271-5G
102523-994EA
87.69
USD
102523-994
102523-992
2-Amino-5-bromobenzothiazole ≥95%
2-Amino-5-bromobenzothiazole
Hazard: Irritant
Formula:
C₇H₅BrN₂S MW: 229.1 g/mol |
MDL Number:
MFCD00577806 CAS Number: 20358-03-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...