6-Bromobenzothiazole ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromobenzo[d]thiazole

072189-5G
102523-558EA 293.85 USD
102523-558
6-Bromobenzothiazole ≥95%
6-Bromobenzothiazole

Hazard: Toxic/Refrigerate

Formula: C₇H₄BrNS
MW: 214.09 g/mol
Melting Pt: 52…54 °C
MDL Number: MFCD04115372
CAS Number: 53218-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR