5-Methylnicotinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Methylpyridine-3-carboxylic acid5-Methyl-3-pyridinecarboxylic acid

072182-5G 072182-100G
102523-524EA 127.69 USD
102523-524 102523-520
5-Methylnicotinic acid ≥95%
5-Methylnicotinic acid

Hazard: Irritant

Formula: C₇H₇NO₂
MW: 137.14 g/mol
MDL Number: MFCD00829036
CAS Number: 3222-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR