4-Fluoro-3-nitrobenzenesulfonamide ≥95%
Supplier: Matrix Scientific
072170-5G
102523-460EA
304.62
USD
102523-460
4-Fluoro-3-nitrobenzenesulfonamide ≥95%
4-Fluoro-3-nitrobenzenesulphonamide
Hazard: Irritant
Formula:
C₆H₅FN₂O₄S MW: 220.18 g/mol |
MDL Number:
MFCD08703185 CAS Number: 406233-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...